Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015621.2 | NP_056436.2 | 9 | 3340 | ||
GenBank mRNA | AK225826.1 | - | 9 | 3310 | ||
AL080169.1 | - | 2 | 2537 | |||
AL713742.1 | - | 8 | 3261 |