Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015658.3 | NP_056473.2 | 13 | 1522 | ||
GenBank mRNA | AK022756.1 | - | 7 | 765 | ||
AK024284.1 | - | 13 | 1476 | |||
AK225239.1 | - | 13 | 1477 | |||
AK315080.1 | - | 13 | 1491 | |||
AL050019.1 | - | 13 | 1478 | |||
BC003555.1 | - | 13 | 1495 | 3546209 |