Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM155B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015686.2 | NP_056501.2 | 1 | 488 | ||
XM_011530908.2 | XP_011529210.1 | 1 | 519 | |||
XM_011530909.2 | XP_011529211.1 | 1 | 519 | |||
GenBank mRNA | AF087142.1 | - | 1 | 487 | ||
BC136518.1 | - | 1 | 475 | 9020508 | ||
BC136519.1 | - | 1 | 475 | 9020509 |