Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128852.1 | NP_001122324.1 | 5 | 804 | ||
NM_001128853.1 | NP_001122325.1 | 5 | 804 | |||
NM_001128854.1 | NP_001122326.1 | 5 | 804 | |||
NM_015908.5 | NP_056992.4 | 5 | 804 | |||
XM_005250405.2 | XP_005250462.1 | 5 | 661 | |||
XM_005250406.2 | XP_005250463.1 | 5 | 661 | |||
XM_005250407.2 | XP_005250464.1 | 5 | 661 | |||
XM_005250408.2 | XP_005250465.1 | 5 | 661 | |||
XM_017012290.1 | XP_016867779.1 | 6 | 986 | |||
XM_017012291.1 | XP_016867780.1 | 6 | 986 | |||
GenBank mRNA | AF248955.1 | - | 5 | 767 | ||
AK302820.1 | - | 5 | 804 | |||
AL096723.1 | - | 5 | 759 | |||
BC069249.1 | - | 3 | 345 | |||
BC109117.1 | - | 5 | 698 | 40035609 | ||
BG477711.1 | - | 187 |