Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM135B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015912.3 | NP_056996.2 | 13 | 2829 | ||
XM_011517056.2 | XP_011515358.1 | 14 | 3635 | |||
XM_011517058.2 | XP_011515360.1 | 13 | 3490 | |||
XM_011517059.1 | XP_011515361.1 | 13 | 2757 | |||
XM_011517060.2 | XP_011515362.1 | 14 | 3617 | |||
XM_011517061.2 | XP_011515363.1 | 14 | 2974 | |||
XM_011517062.1 | XP_011515364.1 | 13 | 2829 | |||
XM_011517063.2 | XP_011515365.1 | 13 | 3472 | |||
XM_011517064.2 | XP_011515366.1 | 14 | 3634 | |||
XM_011517065.1 | XP_011515367.1 | 13 | 2893 | |||
XM_011517068.2 | XP_011515370.1 | 13 | 2722 | |||
XM_011517069.2 | XP_011515371.1 | 8 | 2816 | |||
XM_011517070.2 | XP_011515372.1 | 14 | 3639 | |||
XM_011517072.2 | XP_011515374.1 | 7 | 2074 | |||
XM_011517073.2 | XP_011515375.1 | 7 | 2078 | |||
XM_011517074.1 | XP_011515376.1 | 6 | 1944 | |||
XM_017013470.1 | XP_016868959.1 | 13 | 3441 | |||
XM_017013471.1 | XP_016868960.1 | 14 | 3578 | |||
GenBank mRNA | AB196635.1 | - | 13 | 2830 | ||
AF131744.1 | - | 1 | 588 | |||
AK091433.1 | - | 13 | 2829 | |||
BC037288.1 | - | 14 | 2864 | 5266568 | ||
BC106912.1 | - | 1 | 395 | |||
BC106913.1 | - | 1 | 395 | |||
CR457147.1 | - | 1 | 258 | |||
CR936831.1 | - | 7 | 2022 |