Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242933.1 | NP_001229862.1 | 7 | 816 | ||
NM_003099.4 | NP_003090.2 | 7 | 816 | |||
NM_148955.3 | NP_683758.1 | 5 | 621 | |||
XM_005254611.2 | XP_005254668.1 | 7 | 792 | |||
GenBank mRNA | AB209013.1 | - | 7 | 623 | ||
AF065483.1 | - | 7 | 686 | |||
AF065484.1 | - | 5 | 491 | |||
AK128179.1 | - | 7 | 708 | |||
AK222561.1 | - | 7 | 700 | |||
AK222793.1 | - | 7 | 689 | |||
AK291752.1 | - | 7 | 700 | |||
AK310618.1 | - | 4 | 457 | |||
BC000357.2 | - | 7 | 696 | 2964409 | ||
BQ935515.1 | - | 7 | 690 | |||
BT006983.1 | - | 7 | 686 | |||
U53225.1 | - | 7 | 731 |