Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042510.1 | NP_001035975.1 | 4 | 606 | ||
NM_001267708.1 | NP_001254637.1 | 5 | 650 | |||
NM_001267709.1 | NP_001254638.1 | 3 | 404 | |||
NM_016096.3 | NP_057180.1 | 3 | 491 | |||
GenBank mRNA | AF125099.1 | - | 3 | 327 | ||
AF275802.1 | - | 3 | 372 | |||
AF275809.1 | - | 3 | 372 | |||
AK027802.1 | - | 3 | 1715 | |||
AK290477.1 | - | 3 | 375 | |||
BC015925.1 | - | 1 | 248 | 3909510 | ||
BC068524.1 | - | 3 | 375 | 5263977 | ||
BG704587.1 | - | 3 | 409 | |||
BI669563.1 | - | 5 | 605 | |||
BI755311.1 | - | 4 | 514 | |||
DB231109.1 | - | 3 | 404 |