Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243351.1 | NP_001230280.1 | 11 | 1212 | ||
NM_016118.4 | NP_057202.3 | 11 | 1212 | |||
XM_005250011.2 | XP_005250068.1 | 11 | 1277 | |||
XM_017012304.1 | XP_016867793.1 | 11 | 1271 | |||
XM_017012305.1 | XP_016867794.1 | 11 | 1271 | |||
XM_017012306.1 | XP_016867795.1 | 11 | 1293 | |||
XM_017012307.1 | XP_016867796.1 | 11 | 1274 | |||
XM_017012308.1 | XP_016867797.1 | 11 | 1271 | |||
XM_017012309.1 | XP_016867798.1 | 11 | 1277 | |||
XM_017012310.1 | XP_016867799.1 | 12 | 1374 | |||
XM_017012311.1 | XP_016867800.1 | 12 | 1278 | |||
XM_017012312.1 | XP_016867801.1 | 12 | 1247 | |||
XM_017012313.1 | XP_016867802.1 | 12 | 1374 | |||
XM_017012314.1 | XP_016867803.1 | 12 | 1250 | |||
GenBank mRNA | AF155099.1 | - | 11 | 1143 | ||
AF300717.1 | - | 11 | 1201 | |||
AF459743.1 | - | 11 | 1160 | |||
AY129295.1 | - | 11 | 1160 | |||
BC046354.2 | - | 10 | 1128 | 6070128 | ||
BG105733.1 | - | 4 | 444 |