Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SARAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284239.1 | NP_001271168.1 | 2 | 384 | ||
NM_016127.5 | NP_057211.4 | 3 | 563 | |||
XM_017013567.1 | XP_016869056.1 | 3 | 813 | |||
XM_017013568.1 | XP_016869057.1 | 3 | 241 | |||
XM_017013569.1 | XP_016869058.1 | 3 | 241 | |||
XM_017013570.1 | XP_016869059.1 | 2 | 323 | |||
GenBank mRNA | AB028926.1 | - | 3 | 423 | ||
AF078855.1 | - | 3 | 391 | |||
AF100748.1 | - | 3 | 320 | |||
AF490252.1 | - | 3 | 304 | |||
AK025927.1 | - | 3 | 473 | |||
AK075340.1 | - | 3 | 421 | |||
AK094386.1 | - | 4 | 404 | |||
AK130057.1 | - | 2 | 171 | |||
AK222632.1 | - | 3 | 320 | |||
AK295360.1 | - | 4 | 368 | |||
AK314659.1 | - | 3 | 446 | |||
AK315956.1 | - | 2 | 141 | |||
AY359104.1 | - | 3 | 488 | |||
BC015012.1 | - | 3 | 392 | 3908758 | ||
JQ348891.1 | - | 3 | 432 | |||
JQ348892.1 | - | 2 | 124 |