Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SARAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284239.1 | NP_001271168.1 | 5 | 1773 | ||
NM_016127.5 | NP_057211.4 | 6 | 1952 | |||
XM_017013569.1 | XP_016869058.1 | 6 | 1630 | |||
GenBank mRNA | AB028926.1 | - | 6 | 1812 | ||
AF100748.1 | - | 6 | 1708 | |||
AK025927.1 | - | 6 | 1862 | |||
AK075340.1 | - | 6 | 1810 | |||
AK094386.1 | - | 7 | 1793 | |||
AK130057.1 | - | 5 | 1560 | |||
AY359104.1 | - | 6 | 1877 | |||
BC015012.1 | - | 6 | 1776 | 3908758 | ||
JQ348891.1 | - | 6 | 1821 |