Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016134.3 | NP_057218.1 | 4 | 912 | ||
XM_017012953.1 | XP_016868442.1 | 4 | 912 | |||
XM_017012954.1 | XP_016868443.1 | 4 | 912 | |||
XM_017012955.1 | XP_016868444.1 | 4 | 912 | |||
GenBank mRNA | AF107834.1 | - | 3 | 755 | ||
AF119386.1 | - | 3 | 726 | |||
AK075132.1 | - | 4 | 830 | |||
AK296013.1 | - | 4 | 532 | |||
AK304008.1 | - | 4 | 308 | |||
AK315447.1 | - | 4 | 829 | |||
BC020689.1 | - | 4 | 868 | 4251802 |