Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UIMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199297.2 | NP_001186226.1 | 16 | 2397 | ||
NM_001199298.1 | NP_001186227.1 | 15 | 2172 | |||
NM_001317961.1 | NP_001304890.1 | 15 | 1785 | |||
NM_016290.4 | NP_057374.3 | 15 | 2186 | |||
XM_005265930.2 | XP_005265987.1 | 15 | 2160 | |||
XM_005265932.2 | XP_005265989.1 | 13 | 1296 | |||
XM_005265933.2 | XP_005265990.1 | 13 | 1408 | |||
XM_005265934.2 | XP_005265991.1 | 15 | 1615 | |||
XM_005265935.2 | XP_005265992.1 | 15 | 1624 | |||
XM_005265936.2 | XP_005265993.1 | 14 | 1414 | |||
XM_006714871.2 | XP_006714934.1 | 15 | 2135 | |||
XM_006714874.2 | XP_006714937.1 | 15 | 1599 | |||
XM_017009574.1 | XP_016865063.1 | 14 | 2051 | |||
XM_017009575.1 | XP_016865064.1 | 16 | 1863 | |||
XM_017009576.1 | XP_016865065.1 | 13 | 1309 | |||
XM_017009577.1 | XP_016865066.1 | 13 | 1361 | |||
XM_017009578.1 | XP_016865067.1 | 14 | 1423 | |||
XM_017009579.1 | XP_016865068.1 | 14 | 1323 | |||
GenBank mRNA | AF113538.1 | - | 11 | 976 | ||
AF349313.1 | - | 15 | 2128 | |||
AK023044.1 | - | 16 | 2174 | |||
AL137375.1 | - | 6 | 504 | |||
BC006078.1 | - | 15 | 2093 | 3659756 | ||
BC032561.1 | - | 15 | 1576 | 5578134 | ||
BX537376.1 | - | 15 | 2186 |