Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUZP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016383.4 | NP_057467.1 | 3, 4 | 378 | ||
GenBank mRNA | AF124430.1 | - | 3, 4 | 348 | ||
AK314765.1 | - | 3, 4 | 367 | |||
BC128134.1 | - | 3, 4 | 340 | 40112435 | ||
BC128135.1 | - | 3, 4 | 339 | |||
DB035084.1 | - | 3, 4 | 379 | |||
DB041564.1 | - | 3, 4 | 370 |