Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256539.3 | NP_001243468.2 | 3 | 644 | ||
NM_001256540.3 | NP_001243469.2 | 3 | 644 | |||
NM_001291305.2 | NP_001278234.1 | 3 | 678 | |||
NM_001291307.2 | NP_001278236.1 | 3 | 644 | |||
NM_001317975.1 | NP_001304904.1 | 2 | 733 | |||
NM_016391.7 | NP_057475.2 | 3 | 644 | |||
NR_134316.1 | 2 | 750 | ||||
NR_134318.1 | 3 | 644 | ||||
GenBank mRNA | AF151019.1 | - | 3 | 640 | ||
AF151875.1 | - | 4 | 712 | |||
AF161460.1 | - | 3 | 318 | |||
AK300883.1 | - | 3 | 323 | |||
AK303281.1 | - | 3 | 315 | |||
AW837047.1 | - | 2 | 399 | |||
AY561704.1 | - | 3 | 256 | |||
BC019331.1 | - | 3 | 479 | 3534019 | ||
BC032424.2 | - | 3 | 287 | 5416724 | ||
BC040106.1 | - | 3 | 304 | 6066208 | ||
BP234677.1 | - | 2 | 404 | |||
BU594763.1 | - | 3 | 318 | |||
CB122385.1 | - | 3 | 304 | |||
CR457169.1 | - | 3 | 256 | |||
HY064071.1 | - | 3 | 327 |