Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080391.1 | NP_001073860.1 | 2, 3 | 256 | ||
NM_001206701.1 | NP_001193630.1 | 2, 3 | 256 | |||
NM_001206702.1 | NP_001193631.1 | 2, 3 | 256 | |||
NM_003113.3 | NP_003104.2 | 2, 3 | 256 | |||
GenBank mRNA | AF056322.1 | - | 2, 3 | 146 | ||
AF255565.1 | - | 2, 3 | 146 | |||
AK304205.1 | - | 2, 3 | 213 | |||
BC011562.1 | - | 2, 3 | 194 | 4560914 | ||
DA562355.1 | - | 2, 3 | 256 | |||
M60618.1 | - | 2, 3 | 146 | |||
U36501.1 | - | 2, 3 | 146 |