Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042452.1 | NP_001035917.1 | 2 | 159 | ||
NM_016542.3 | NP_057626.2 | 3 | 450 | |||
XM_011531349.1 | XP_011529651.1 | 4 | 423 | |||
XM_011531350.1 | XP_011529652.1 | 3 | 365 | |||
GenBank mRNA | AB040057.2 | - | 3 | 369 | ||
AF231012.1 | - | 3 | 338 | |||
AF344882.1 | - | 2 | 159 | |||
AF344883.1 | - | 2 | 159 | |||
AK075150.1 | - | 3 | 365 | |||
AK303583.1 | - | 3 | 365 | |||
AK314356.1 | - | 3 | 366 | |||
AK316137.1 | - | 3 | 365 | |||
BC098315.1 | - | 3 | 355 | 40026349 | ||
BC099843.1 | - | 3 | 355 | 40026348 | ||
BC103503.1 | - | 3 | 355 | 40026350 | ||
BT020099.1 | - | 2 | 149 |