Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RXFP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016568.3 | NP_057652.1 | 1 | 743 | ||
GenBank mRNA | AY394501.1 | - | 1 | 388 | ||
BC095526.1 | - | 1 | 462 | 30706560 | ||
BC113438.1 | - | 1 | 464 | 8322490 | ||
BC113440.1 | - | 1 | 464 | 8322492 | ||
D88437.1 | - | 1 | 748 |
Set Membership: |
G-protein coupled receptor |