Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317812.1 | NP_001304741.1 | 3 | 1164 | ||
NM_001317813.1 | NP_001304742.1 | 4 | 811 | |||
NM_001317814.1 | NP_001304743.1 | 4 | 758 | |||
NM_016612.3 | NP_057696.2 | 3 | 687 | |||
XM_006716352.3 | XP_006716415.1 | 3 | 291 | |||
XM_011544550.2 | XP_011542852.1 | 3 | 291 | |||
XM_011544553.2 | XP_011542855.1 | 4 | 1523 | |||
XM_011544554.2 | XP_011542856.1 | 3 | 2445 | |||
XM_017013532.1 | XP_016869021.1 | 3 | 1362 | |||
GenBank mRNA | AF113696.1 | - | 1 | 488 | ||
AF116630.1 | - | 1 | 1310 | |||
AF155660.1 | - | 1 | 505 | |||
AF223466.1 | - | 3 | 523 | |||
AK074708.1 | - | 3 | 1003 | |||
AK075313.1 | - | 3 | 928 | |||
AK127666.1 | - | 3 | 2756 | |||
AK223194.1 | - | 3 | 1004 | |||
AL833186.1 | - | 2 | 2406 | |||
AY032628.1 | - | 2 | 2510 | |||
BC132799.1 | - | 3 | 551 | 40146821 | ||
BC132801.1 | - | 3 | 551 | 40146823 | ||
BI460337.1 | - | 3 | 641 | |||
BM552840.1 | - | 4 | 693 | |||
BM926580.1 | - | 3 | 530 | |||
BU166030.1 | - | 4 | 731 |