Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300783.1 | NP_001287712.1 | 2 | 907 | ||
NM_001308087.1 | NP_001295016.1 | 2 | 620 | |||
NM_001308088.1 | NP_001295017.1 | 4 | 860 | |||
NM_016644.2 | NP_057728.1 | 3 | 1032 | |||
XM_011543452.2 | XP_011541754.1 | 3 | 2519 | |||
XM_011543453.2 | XP_011541755.1 | 3 | 1001 | |||
XM_011543454.2 | XP_011541756.1 | 4 | 1107 | |||
GenBank mRNA | AF242769.1 | - | 3 | 986 | ||
BC038838.1 | - | 3 | 719 | 5170587 | ||
BC092474.1 | - | 2 | 908 | 30528467 |