Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317931.1 | NP_001304860.1 | 6 | 700 | ||
NM_003116.2 | NP_003107.1 | 6 | 719 | |||
XM_005260520.4 | XP_005260577.1 | 6 | 365 | |||
XM_011529009.2 | XP_011527311.1 | 6 | 615 | |||
GenBank mRNA | AF262992.1 | - | 6 | 716 | ||
BC143783.1 | - | 6 | 621 |