Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017621.3 | NP_060091.1 | 3 | 403 | ||
XM_005250464.2 | XP_005250521.1 | 3 | 296 | |||
XM_005250465.2 | XP_005250522.1 | 3 | 382 | |||
GenBank mRNA | AK000020.1 | - | 3 | 390 | ||
AK026097.1 | - | 3 | 301 | |||
AK222689.1 | - | 3 | 392 | |||
BC002820.2 | - | 3 | 370 | 3636984 | ||
BC017096.1 | - | 3 | 370 | 2820440 | ||
BX353099.2 | - | 3 | 404 |