Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TET2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127208.2 | NP_001120680.1 | 3 | 3248 | ||
NM_017628.4 | NP_060098.3 | 3 | 3165 | |||
XM_005263082.2 | XP_005263139.1 | 2 | 3124 | |||
XM_006714242.3 | XP_006714305.1 | 3 | 3092 | |||
XM_017008319.1 | XP_016863808.1 | 3 | 3092 | |||
GenBank mRNA | AB075496.1 | - | 1 | 393 | ||
AK027819.1 | - | 1 | 948 | |||
BC150180.1 | - | 2 | 2863 | |||
BC150201.1 | - | 2 | 2863 | |||
BX640738.1 | - | 3 | 3166 | |||
FM992369.1 | - | 3 | 3621 |