Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX60L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012967.2 | NP_001012985.2 | 4 | 494 | ||
NM_001291510.1 | NP_001278439.1 | 4 | 494 | |||
XM_005263341.4 | XP_005263398.2 | 4 | 477 | |||
XM_005263342.2 | XP_005263399.2 | 4 | 494 | |||
XM_005263343.2 | XP_005263400.2 | 4 | 460 | |||
XM_005263345.4 | XP_005263402.2 | 4 | 473 | |||
XM_006714408.2 | XP_006714471.1 | 5 | 544 | |||
XM_011532402.2 | XP_011530704.1 | 5 | 767 | |||
XM_011532403.2 | XP_011530705.1 | 5 | 602 | |||
XM_011532404.1 | XP_011530706.1 | 4 | 494 | |||
XM_011532405.1 | XP_011530707.1 | 4 | 494 | |||
XM_011532406.1 | XP_011530708.1 | 4 | 494 | |||
XM_017008829.1 | XP_016864318.1 | 4 | 460 | |||
XM_017008830.1 | XP_016864319.1 | 4 | 494 | |||
GenBank mRNA | AK096369.1 | - | 5 | 544 | ||
CR933601.1 | - | 4 | 468 | |||
DA553350.1 | - | 4 | 494 | |||
KR632542.1 | - | 2 | 246 |
Gene Symbol: DDX60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017631.5 | NP_060101.3 | 4 | 538 | ||
XM_011532103.2 | XP_011530405.1 | 4 | 538 | |||
XM_011532104.2 | XP_011530406.1 | 4 | 538 | |||
XM_017008383.1 | XP_016863872.1 | 4 | 538 | |||
XM_017008384.1 | XP_016863873.1 | 4 | 538 | |||
GenBank mRNA | AK001649.1 | - | 4 | 414 | ||
AK307404.1 | - | 4 | 538 | |||
BC038115.2 | - | 4 | 483 | 5261001 |