Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017631.5 | NP_060101.3 | 14, 15 | 2278 | ||
XM_011532103.2 | XP_011530405.1 | 14, 15 | 2278 | |||
XM_011532104.2 | XP_011530406.1 | 14, 15 | 2278 | |||
XM_017008383.1 | XP_016863872.1 | 14, 15 | 2278 | |||
XM_017008384.1 | XP_016863873.1 | 14, 15 | 2278 | |||
GenBank mRNA | AK001649.1 | - | 14, 15 | 2154 | ||
AK307404.1 | - | 14, 15 | 2278 | |||
BC038115.2 | - | 14, 15 | 2223 | 5261001 |