Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN2AIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017632.3 | NP_060102.1 | 2 | 600 | ||
GenBank mRNA | AF246705.1 | - | 2 | 548 | ||
AK000043.1 | - | 2 | 555 | |||
AK225848.1 | - | 2 | 413 | |||
BC022270.1 | - | 2 | 437 | 4697255 | ||
CN427056.1 | - | 2 | 601 |