Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193307.1 | NP_001180236.1 | 2 | 489 | ||
NM_017654.3 | NP_060124.2 | 3 | 590 | |||
GenBank mRNA | AB095925.2 | - | 3 | 524 | ||
AF445355.1 | - | 3 | 590 | |||
AK122951.1 | - | 2 | 428 | |||
BC065769.1 | - | 3 | 580 | |||
BC132773.1 | - | 2 | 420 | 40146795 | ||
BC132775.1 | - | 2 | 420 | 40146797 | ||
BC150249.1 | - | 3 | 545 | 9007474 | ||
BG615970.1 | - | 3 | 577 | |||
BX647072.1 | - | 3 | 546 |