Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127178.2 | NP_001120650.1 | 3, 4 | 714 | ||
NM_001289051.1 | NP_001275980.1 | 3, 4 | 523 | |||
NM_001289053.1 | NP_001275982.1 | 3, 4 | 1272 | |||
NM_001289055.1 | NP_001275984.1 | 4, 5 | 784 | |||
NM_001289057.1 | NP_001275986.1 | 3, 4 | 950 | |||
NM_017733.4 | NP_060203.3 | 3, 4 | 714 | |||
NR_110293.1 | 3, 4 | 714 | ||||
XM_005272284.2 | XP_005272341.1 | 3, 4 | 948 | |||
XM_011513490.2 | XP_011511792.1 | 3, 4 | 677 | |||
XM_011513491.1 | XP_011511793.1 | 4, 5 | 744 | |||
XM_011513493.1 | XP_011511795.1 | 3, 4 | 714 | |||
XM_011513494.2 | XP_011511796.1 | 3, 4 | 714 | |||
XM_017008321.1 | XP_016863810.1 | 3, 4 | 948 | |||
XM_017008322.1 | XP_016863811.1 | 3, 4 | 948 | |||
XM_017008323.1 | XP_016863812.1 | 3, 4 | 948 | |||
XM_017008324.1 | XP_016863813.1 | 3, 4 | 948 | |||
XM_017008325.1 | XP_016863814.1 | 3, 4 | 661 | |||
XM_017008326.1 | XP_016863815.1 | 3, 4 | 714 | |||
GenBank mRNA | AB162713.1 | - | 3, 4 | 684 | ||
AK074715.1 | - | 3, 4 | 486 | |||
AK074815.1 | - | 3, 4 | 693 | |||
AK097244.1 | - | 3, 4 | 911 | |||
AK296507.1 | - | 4, 5 | 775 | |||
AY358538.1 | - | 3, 4 | 656 | |||
BC050262.1 | - | 3, 4 | 1184 | |||
BC144576.1 | - | 3, 4 | 671 |