Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017752.2 | NP_060222.2 | 4 | 655 | ||
NM_198881.1 | NP_942582.1 | 4 | 655 | |||
XM_006724669.1 | XP_006724732.1 | 4 | 655 | |||
XM_006724670.3 | XP_006724733.1 | 4 | 469 | |||
GenBank mRNA | AB449890.1 | - | 4 | 490 | ||
AB449891.1 | - | 4 | 490 | |||
AK123957.1 | - | 4 | 655 | |||
BC122564.1 | - | 4 | 654 | 40026331 | ||
BX648230.1 | - | 4 | 639 |