Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEMIN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042479.1 | NP_001035944.1 | 4 | 564 | ||
NM_001042480.1 | NP_001035945.1 | 3 | 482 | |||
NM_017856.2 | NP_060326.1 | 4 | 652 | |||
XM_005274555.2 | XP_005274612.1 | 3 | 3517 | |||
XM_017029617.1 | XP_016885106.1 | 3 | 653 | |||
XM_017029618.1 | XP_016885107.1 | 3 | 653 | |||
GenBank mRNA | AK000521.1 | - | 4 | 629 | ||
AK125062.1 | - | 3 | 2609 | |||
BC003607.2 | - | 3 | 406 | 2987964 | ||
BC014324.1 | - | 3 | 463 | 3930643 | ||
BC020785.1 | - | 3 | 459 | 4808527 | ||
BX386466.2 | - | 4 | 530 | |||
CR457241.1 | - | 1 | 309 | |||
DQ224033.1 | - | 1 | 309 |