Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URGCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077663.2 | NP_001071131.1 | 6 | 3799 | ||
NM_001077664.2 | NP_001071132.1 | 6 | 3571 | |||
NM_001290075.1 | NP_001277004.1 | 6 | 3794 | |||
NM_001290076.1 | NP_001277005.1 | 7 | 3693 | |||
NM_017920.4 | NP_060390.3 | 5 | 3772 | |||
GenBank mRNA | AB040940.1 | - | 7 | 4096 | ||
AK000661.1 | - | 1 | 1506 | |||
AK026185.1 | - | 1 | 2978 | |||
AL832134.1 | - | 4 | 5412 | |||
AL833744.1 | - | 1 | 1902 | |||
AY078404.1 | - | 5 | 3293 | |||
BC018426.1 | - | 5 | 3295 | 4213413 | ||
BX640797.1 | - | 6 | 3334 | |||
BX648505.1 | - | 6 | 3341 |