Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166373.1 | NP_001159845.1 | 8 | 1240 | ||
NM_017923.3 | NP_060393.1 | 4 | 1127 | |||
XM_011532055.2 | XP_011530357.1 | 8 | 1945 | |||
XM_017008334.1 | XP_016863823.1 | 7 | 1167 | |||
XM_017008335.1 | XP_016863824.1 | 8 | 1546 | |||
XM_017008336.1 | XP_016863825.1 | 8 | 1412 | |||
GenBank mRNA | AK000675.1 | - | 4 | 1016 | ||
AK289711.1 | - | 8 | 1240 | |||
AL713759.1 | - | 8 | 2039 |