Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXorf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184782.1 | NP_001171711.1 | 6 | 1402 | ||
NM_018015.5 | NP_060485.4 | 6 | 1402 | |||
GenBank mRNA | AK001040.1 | - | 6 | 1357 | ||
AK024253.1 | - | 6 | 676 | |||
AK291730.1 | - | 6 | 1351 | |||
BC011483.1 | - | 6 | 1323 | 3453700 | ||
BC033709.1 | - | 6 | 1314 | 3910777 | ||
BC070110.1 | - | 6 | 1353 | 5264812 |