Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018051.4 | NP_060521.4 | 17 | 2358 | ||
XM_005249549.1 | XP_005249606.1 | 14 | 1950 | |||
XM_006716041.2 | XP_006716104.1 | 17 | 2382 | |||
XM_011516367.1 | XP_011514669.1 | 17 | 2224 | |||
XM_011516368.2 | XP_011514670.1 | 17 | 2382 | |||
XM_011516369.2 | XP_011514671.1 | 17 | 2213 | |||
XM_017012381.1 | XP_016867870.1 | 17 | 2404 | |||
XM_017012382.1 | XP_016867871.1 | 17 | 2373 | |||
XM_017012383.1 | XP_016867872.1 | 17 | 2434 | |||
XM_017012384.1 | XP_016867873.1 | 14 | 1941 | |||
XM_017012385.1 | XP_016867874.1 | 16 | 2115 | |||
XM_017012386.1 | XP_016867875.1 | 14 | 1894 | |||
XM_017012387.1 | XP_016867876.1 | 17 | 2234 | |||
XM_017012388.1 | XP_016867877.1 | 7 | 1118 | |||
GenBank mRNA | AK001162.1 | - | 1 | 73 | ||
AK027228.1 | - | 6 | 715 | |||
AL137660.1 | - | 12 | 1358 | |||
BC014491.1 | - | 17 | 2358 | 4906683 | ||
BM718580.1 | - | 2 | 181 | |||
CR627036.1 | - | 12 | 4000 |