Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256891.1 | NP_001243820.1 | 2 | 162 | ||
NM_001256892.1 | NP_001243821.1 | 2 | 138 | |||
NM_003130.3 | NP_003121.1 | 2 | 162 | |||
NM_198901.1 | NP_944490.1 | 2 | 138 | |||
GenBank mRNA | AK129770.1 | - | 2 | 131 | ||
AK295225.1 | - | 2 | 115 | |||
AK296601.1 | - | 2 | 121 | |||
AK310366.1 | - | 2 | 138 | |||
BC011025.1 | - | 2 | 120 | 4281626 | ||
BI524104.1 | - | 2 | 122 | |||
BI559592.1 | - | 2 | 156 | |||
BI597920.1 | - | 2 | 158 | |||
BM739144.1 | - | 2 | 138 | |||
DA268050.1 | - | 2 | 162 | |||
L12387.1 | - | 2 | 108 | |||
M32886.1 | - | 2 | 116 |