Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166135.1 | NP_001159607.1 | 4 | 573 | ||
NM_018077.2 | NP_060547.2 | 8 | 996 | |||
XM_011516370.2 | XP_011514672.1 | 9 | 1103 | |||
XM_017012389.1 | XP_016867878.1 | 9 | 1104 | |||
XM_017012390.1 | XP_016867879.1 | 8 | 996 | |||
GenBank mRNA | AK001239.1 | - | 8 | 996 | ||
AK222716.1 | - | 8 | 958 | |||
AK300500.1 | - | 4 | 535 | |||
BC013889.2 | - | 8 | 950 | 3926178 | ||
KU178685.1 | - | 8 | 881 |