Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018140.3 | NP_060610.2 | 7 | 1082 | ||
XM_005248322.3 | XP_005248379.1 | 7 | 1004 | |||
XM_011514063.1 | XP_011512365.1 | 7 | 1028 | |||
XM_011514064.2 | XP_011512366.1 | 7 | 1135 | |||
XM_017009626.1 | XP_016865115.1 | 6 | 916 | |||
XM_017009627.1 | XP_016865116.1 | 6 | 876 | |||
GenBank mRNA | AB040952.1 | - | 7 | 1020 | ||
AK001427.1 | - | 7 | 1025 | |||
AK299072.1 | - | 6 | 836 | |||
BC000132.2 | - | 7 | 1021 | 2899944 | ||
BC001750.1 | - | 7 | 1008 |