Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047701.1 | 8 | 1226 | |||
NR_135583.1 | 7 | 1161 | ||||
XM_005249807.4 | XP_005249864.1 | 7 | 1225 | |||
XM_006715752.2 | XP_006715815.1 | 8 | 1193 | |||
XM_017012414.1 | XP_016867903.1 | 7 | 1152 | |||
XM_017012415.1 | XP_016867904.1 | 8 | 1254 | |||
XM_017012416.1 | XP_016867905.1 | 7 | 1152 | |||
XM_017012421.1 | XP_016867910.1 | 8 | 1376 | |||
XM_017012422.1 | XP_016867911.1 | 8 | 1265 | |||
XM_017012424.1 | XP_016867913.1 | 7 | 1168 | |||
GenBank mRNA | AK001665.1 | - | 8 | 1196 | ||
AK023007.1 | - | 8 | 1179 |