Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB061546.1 | - | 3 | 211 | ||
AB451391.1 | - | 3 | 138 | |||
BC071716.1 | - | 3 | 160 | 4476669 | ||
BC100031.1 | - | 3 | 164 | 2847600 | ||
DA338824.1 | - | 3 | 224 | |||
X73459.1 | - | 3 | 157 |