Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304529.1 | NP_001291458.1 | 7 | 2828 | ||
NM_001304530.1 | NP_001291459.1 | 7 | 3037 | |||
NM_001304532.1 | NP_001291461.1 | 8 | 3083 | |||
NM_018246.2 | NP_060716.2 | 9 | 3135 | |||
NR_130761.1 | 7 | 2985 | ||||
NR_130762.1 | 7 | 2979 | ||||
XM_011544570.2 | XP_011542872.1 | 8 | 3082 | |||
XM_011544571.2 | XP_011542873.1 | 7 | 2961 | |||
GenBank mRNA | AK001715.1 | - | 8 | 3095 | ||
BC065295.1 | - | 9 | 3089 | 6139413 |