Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324242.1 | NP_001311171.1 | 8 | 1592 | ||
NM_001324243.1 | NP_001311172.1 | 8 | 1685 | |||
NM_018301.4 | NP_060771.3 | 7 | 1520 | |||
NR_136728.1 | 7 | 1188 | ||||
NR_136729.1 | 8 | 1282 | ||||
NR_136730.1 | 8 | 2316 | ||||
NR_136732.1 | 7 | 1281 | ||||
XM_011530981.2 | XP_011529283.1 | 7 | 1612 | |||
GenBank mRNA | AK023732.1 | - | 8 | 2311 | ||
BC006986.1 | - | 7 | 1497 | 3995903 |