Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRIX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018321.3 | NP_060791.3 | 3, 4 | 344 | ||
GenBank mRNA | AF347667.1 | - | 3, 4 | 320 | ||
AK001962.1 | - | 3, 4 | 349 | |||
AK289610.1 | - | 3, 4 | 343 | |||
AK303311.1 | - | 3, 4 | 383 | |||
AY364243.1 | - | 3, 4 | 320 | |||
AY364244.1 | - | 3, 4 | 320 | |||
BC001546.1 | - | 3, 4 | 344 | 3462041 | ||
BC006014.1 | - | 3, 4 | 347 | |||
BC036741.1 | - | 3, 4 | 694 | 5107954 |