Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018390.3 | NP_060860.1 | 7 | 4927 | ||
NR_028057.1 | 9 | 1710 | ||||
XM_006724866.2 | XP_006724929.1 | 7 | 4679 | |||
GenBank mRNA | AK002185.1 | - | 10 | 1712 | ||
AK091006.1 | - | 1 | 2108 | |||
AK097346.1 | - | 1 | 1365 | |||
BC005028.1 | - | 8 | 1487 | 3953383 | ||
BC064136.1 | - | 3 | 549 |