Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ALG13 SIRNA Assays ›
Gene Symbol:
ALG13
Gene Name:
ALG13, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases:
CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W
Chromosome Location:
Chr.X: 111680741 - 111764164 on Build GRCh38
UniGene ID:
Hs.443061
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
127167
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ALG13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039210.4
NP_001034299.3
3
332
NM_001099922.2
NP_001093392.1
3
391
NM_001168385.2
NP_001161857.1
3
391
NM_001257230.1
NP_001244159.1
3
287
NM_001257231.1
NP_001244160.1
3
405
NM_001257234.1
NP_001244163.1
3
360
NM_001257235.2
NP_001244164.1
3
360
NM_001257237.1
NP_001244166.1
3
464
NM_001257239.2
NP_001244168.1
3
379
NM_001257240.2
NP_001244169.1
3
464
NM_001257241.2
NP_001244170.1
3
405
NM_001324290.1
NP_001311219.1
3
397
NM_001324291.1
NP_001311220.1
3
287
NM_001324292.1
NP_001311221.1
3
391
NM_001324293.1
NP_001311222.1
3
464
NM_001324294.1
NP_001311223.1
3
275
NM_018466.5
NP_060936.1
3
391
NR_033125.2
3
287
XM_006724693.2
XP_006724756.1
3
764
XM_006724697.3
XP_006724760.2
3
664
XM_006724698.3
XP_006724761.2
3
789
XM_011531028.2
XP_011529330.1
3
765
XM_011531030.1
XP_011529332.1
3
763
XM_011531031.1
XP_011529333.1
3
464
XM_011531032.1
XP_011529334.1
3
270
XM_011531033.2
XP_011529335.1
3
310
XM_017029846.1
XP_016885335.1
3
766
XM_017029847.1
XP_016885336.1
3
765
XM_017029848.1
XP_016885337.1
3
763
XM_017029849.1
XP_016885338.1
3
764
XM_017029850.1
XP_016885339.1
3
763
XM_017029851.1
XP_016885340.1
3
766
GenBank mRNA
AF220051.1
-
3
322
AK300394.1
-
3
229
AK302729.1
-
3
332
AK302890.1
-
3
308
AK304712.1
-
3
281
AK311608.1
-
3
271
AK312229.1
-
3
330
AK316306.1
-
3
333
AK316522.1
-
3
408
AL524204.3
-
3
210
BC005336.1
-
3
324
3996922
BE747995.1
-
3
320
BF239206.1
-
3
359
BF240663.1
-
3
177
BF699152.1
-
3
393
BG621042.1
-
2
174
BM759764.1
-
3
373
CB268871.1
-
3
338
DA830058.1
-
3
455
DB292609.1
-
3
464
DC400909.1
-
3
275
DN992019.1
-
3
459
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
transferase
Gene Ontology Categories:
Function(s)
Process(es)
dolichol-linked oligosaccharide biosynthetic process
proteolysis
N-acetylglucosaminyldiphosphodolichol N-acetylglucosaminyltransferase activity
thiol-dependent ubiquitinyl hydrolase activity
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-vpmkj:80/100.66.78.144:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline