Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1AR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018569.5 | NP_061039.3 | 6 | 683 | ||
NR_136622.1 | 6 | 683 | ||||
XM_017008378.1 | XP_016863867.1 | 7 | 842 | |||
XM_017008380.1 | XP_016863869.1 | 6 | 366 | |||
GenBank mRNA | AK310787.1 | - | 6 | 682 | ||
AK315299.1 | - | 6 | 583 | |||
AL136628.1 | - | 6 | 601 | |||
BM823928.1 | - | 6 | 295 | |||
BX647702.1 | - | 6 | 636 |