Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAGE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018666.2 | NP_061136.2 | 4 | 429 | ||
XM_017029621.1 | XP_016885110.1 | 4 | 437 | |||
XM_017029622.1 | XP_016885111.1 | 4 | 348 | |||
XM_017029623.1 | XP_016885112.1 | 4 | 344 | |||
XM_017029625.1 | XP_016885114.1 | 4 | 344 | |||
GenBank mRNA | AJ278111.1 | - | 4 | 429 | ||
BC144260.1 | - | 4 | 310 | 9052780 |