Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC4H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178032.2 | NP_001171503.1 | 3 | 632 | ||
NM_001178033.2 | NP_001171504.1 | 3 | 470 | |||
NM_001243804.1 | NP_001230733.1 | 3 | 377 | |||
NM_018684.3 | NP_061154.1 | 3 | 470 | |||
NR_045044.1 | 3 | 725 | ||||
GenBank mRNA | AB032992.1 | - | 3 | 470 | ||
AF270491.1 | - | 3 | 402 | |||
AK022807.1 | - | 3 | 601 | |||
AK022918.1 | - | 3 | 364 | |||
AK123763.1 | - | 4 | 863 | |||
AK222708.1 | - | 3 | 387 | |||
AK293570.1 | - | 3 | 387 | |||
AK315485.1 | - | 3 | 385 | |||
BC004411.1 | - | 3 | 421 | 3639978 | ||
BP376634.1 | - | 3 | 377 | |||
CX782941.1 | - | 3 | 342 |