Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM134B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034850.2 | NP_001030022.1 | 7, 8 | 954 | ||
NM_019000.4 | NP_061873.2 | 5, 6 | 830 | |||
XM_011514053.2 | XP_011512355.1 | 8, 9 | 1013 | |||
XM_011514054.2 | XP_011512356.1 | 6, 7 | 964 | |||
XM_011514055.2 | XP_011512357.1 | 5, 6 | 754 | |||
GenBank mRNA | AK024920.1 | - | 5, 6 | 660 | ||
AK025808.1 | - | 7, 8 | 881 | |||
AL832438.1 | - | 7, 8 | 630 | |||
BC030517.1 | - | 1, 2 | 26 | |||
BC053326.1 | - | 7, 8 | 878 | 5014868 | ||
BC073132.1 | - | 5, 6 | 663 | 5760145 | ||
CR994722.1 | - | 1, 2 | 69 |