Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMCX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009584.1 | NP_001009584.1 | 2 | 1176 | ||
NM_001184768.1 | NP_001171697.1 | 4 | 1346 | |||
NM_019007.3 | NP_061880.2 | 3 | 1258 | |||
NR_033669.1 | 4 | 347 | ||||
NR_033670.1 | 5 | 435 | ||||
XM_011530976.2 | XP_011529278.1 | 4 | 1312 | |||
GenBank mRNA | AK000818.1 | - | 1 | 840 | ||
AK097018.1 | - | 1 | 1650 | |||
AW955663.1 | - | 5 | 347 | |||
BC007677.2 | - | 3 | 1258 | 3609980 | ||
BX423052.2 | - | 4 | 348 | |||
L20773.1 | - | 1 | 976 |