Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318163.1 | NP_001305092.1 | 12 | 1500 | ||
NM_001318164.1 | NP_001305093.1 | 12 | 1689 | |||
NM_019042.4 | NP_061915.2 | 12 | 1713 | |||
XM_005250462.3 | XP_005250519.1 | 13 | 1739 | |||
XM_011516336.2 | XP_011514638.1 | 13 | 1734 | |||
XM_017012367.1 | XP_016867856.1 | 13 | 1667 | |||
XM_017012368.1 | XP_016867857.1 | 11 | 1580 | |||
GenBank mRNA | AB067484.1 | - | 12 | 1427 | ||
AK000492.1 | - | 2 | 217 | |||
AK091283.1 | - | 12 | 1693 | |||
AK128629.1 | - | 12 | 1675 | |||
BC005209.2 | - | 4 | 426 | |||
BC011396.1 | - | 12 | 1669 | 3870711 |