Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184965.1 | NP_001171894.1 | 4 | 934 | ||
NM_001184966.1 | NP_001171895.1 | 3 | 859 | |||
NM_019045.4 | NP_061918.3 | 4 | 934 | |||
XM_011531353.2 | XP_011529655.1 | 3 | 935 | |||
XM_017029600.1 | XP_016885089.1 | 3 | 867 | |||
GenBank mRNA | AK127556.1 | - | 4 | 928 | ||
AK299711.1 | - | 3 | 795 | |||
AK304432.1 | - | 4 | 461 | |||
AL833605.1 | - | 4 | 899 | |||
AL834210.1 | - | 1 | 201 | |||
BC028697.2 | - | 4 | 870 | 4837674 | ||
BX537578.1 | - | 4 | 909 | |||
BX538087.1 | - | 4 | 876 | |||
BX648150.1 | - | 4 | 873 |